siRNA Details
| Gene Symbol: Gpr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025381.2 | NP_001020552.1 | 2 | 2306 | ||
| GenBank mRNA | BC070439.1 | - | 2 | 2306 | 30533246 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr17| Gene Symbol: Gpr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025381.2 | NP_001020552.1 | 2 | 2306 | ||
| GenBank mRNA | BC070439.1 | - | 2 | 2306 | 30533246 | |