siRNA Details
| Gene Symbol: G930045G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145307.1 | - | 5 | 890 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G930045G22Rik| Gene Symbol: G930045G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145307.1 | - | 5 | 890 | ||