siRNA Details
| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026281.2 | NP_080557.2 | 2 | 303 | ||
| GenBank mRNA | AK010720.1 | - | 2 | 146 | ||
| AK154633.1 | - | 2 | 303 | |||
| AK171111.1 | - | 2 | 280 | |||
| AK204708.1 | - | 2 | 132 | |||
| BC023389.1 | - | 2 | 241 | |||
| BC049960.1 | - | 2 | 211 | |||
| DQ104329.1 | - | 2 | 251 | |||

Tm7sf3
Genomic Map