siRNA Details
| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK171111.1 | - | 2 | 328 | ||
| BC023389.1 | - | 2 | 289 | |||
| BC049960.1 | - | 2 | 259 | |||
| DQ104329.1 | - | 2 | 299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm7sf3| Gene Symbol: Tm7sf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK171111.1 | - | 2 | 328 | ||
| BC023389.1 | - | 2 | 289 | |||
| BC049960.1 | - | 2 | 259 | |||
| DQ104329.1 | - | 2 | 299 | |||