siRNA Details
| Gene Symbol: FRAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005479.3 | NP_005470.2 | 1 | 1208 | ||
| GenBank mRNA | AB074890.1 | - | 1 | 1132 | ||
| U58975.2 | - | 1 | 1208 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FRAT1| Gene Symbol: FRAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005479.3 | NP_005470.2 | 1 | 1208 | ||
| GenBank mRNA | AB074890.1 | - | 1 | 1132 | ||
| U58975.2 | - | 1 | 1208 | |||