siRNA Details
| Gene Symbol: Tmem129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026698.2 | NP_080974.2 | 4 | 1695 | ||
| GenBank mRNA | AK010497.1 | - | 1 | 505 | ||
| AK172044.1 | - | 4 | 1824 | |||
| BC094042.1 | - | 4 | 1695 | 5098723 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem129| Gene Symbol: Tmem129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026698.2 | NP_080974.2 | 4 | 1695 | ||
| GenBank mRNA | AK010497.1 | - | 1 | 505 | ||
| AK172044.1 | - | 4 | 1824 | |||
| BC094042.1 | - | 4 | 1695 | 5098723 | ||