siRNA Details
Gene Symbol: Ecscr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033141.1 | NP_001028313.1 | 9 | 1034 | ||
GenBank mRNA | AK003507.1 | - | 5 | 527 | ||
AK157485.1 | - | 9 | 1034 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ecscr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033141.1 | NP_001028313.1 | 9 | 1034 | ||
GenBank mRNA | AK003507.1 | - | 5 | 527 | ||
AK157485.1 | - | 9 | 1034 |