siRNA Details
| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026805.1 | NP_081081.1 | 16 | 2556 | ||
| GenBank mRNA | AK003981.1 | - | 1 | 831 | ||
| AK053833.1 | - | 16 | 2547 | |||
| BC059878.1 | - | 16 | 2509 | 6812082 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Svop| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026805.1 | NP_081081.1 | 16 | 2556 | ||
| GenBank mRNA | AK003981.1 | - | 1 | 831 | ||
| AK053833.1 | - | 16 | 2547 | |||
| BC059878.1 | - | 16 | 2509 | 6812082 | ||