siRNA Details
| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026805.1 | NP_081081.1 | 15, 16 | 1642 | ||
| GenBank mRNA | AK029670.1 | - | 14, 15 | 1351 | ||
| AK047655.1 | - | 15, 16 | 1635 | |||
| AK047764.1 | - | 15, 16 | 1643 | |||
| AK053833.1 | - | 15, 16 | 1633 | |||
| AK082744.1 | - | 15, 16 | 1644 | |||
| BC059878.1 | - | 15, 16 | 1595 | 6812082 | ||

Svop
Genomic Map