siRNA Details
| Gene Symbol: Tyw5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037742.2 | NP_001032831.2 | 8 | 974 | ||
| GenBank mRNA | BC023868.1 | - | 8 | 966 | ||
| BC058590.1 | - | 7 | 1290 | |||
| BC099495.1 | - | 8 | 716 | |||
| BC132289.1 | - | 8 | 823 | 40130566 | ||
| BC144765.1 | - | 8 | 789 | |||
| BC144766.1 | - | 8 | 804 | |||

Tyw5
Genomic Map