siRNA Details
| Gene Symbol: Tyw5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037742.2 | NP_001032831.2 | 6, 7 | 795 | ||
| GenBank mRNA | BC023868.1 | - | 6, 7 | 787 | ||
| BC058590.1 | - | 5, 6 | 1111 | |||
| BC099495.1 | - | 6, 7 | 537 | |||
| BC132289.1 | - | 6, 7 | 644 | 40130566 | ||
| BC144766.1 | - | 6, 7 | 625 | |||
| CF951173.1 | - | 7, 8 | 571 | |||

Tyw5
Genomic Map