siRNA Details
| Gene Symbol: Mybphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026831.1 | NP_081107.1 | 2 | 277 | ||
| GenBank mRNA | AK004148.1 | - | 2 | 278 | ||
| BC089625.1 | - | 2 | 265 | 6756382 | ||
| BC132636.1 | - | 2 | 266 | 40130913 | ||
| BC132638.1 | - | 2 | 266 | 40130915 | ||
| BY704393.1 | - | 2 | 278 | |||

Mybphl
Genomic Map