siRNA Details
| Gene Symbol: Psmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027357.2 | NP_081633.1 | 18, 19 | 2284 | ||
| GenBank mRNA | AK010596.1 | - | 2, 3 | 230 | ||
| AK087947.1 | - | 18, 19 | 2235 | |||
| AK153386.1 | - | 18, 19 | 2285 | |||
| AK159123.1 | - | 18, 19 | 2284 | |||
| AK159750.1 | - | 18, 19 | 2263 | |||
| BC018172.1 | - | 4, 5 | 333 | |||
| BC022955.1 | - | 18, 19 | 2174 | |||
| BC025802.1 | - | 18, 19 | 2188 | |||
| BC029319.1 | - | 18, 19 | 2221 | |||
| BC031587.1 | - | 18, 19 | 2215 | |||
| BC032068.1 | - | 18, 19 | 2215 | |||
| BC039937.1 | - | 18, 19 | 2261 | |||
| BC089028.1 | - | 18, 19 | 2231 | |||
| BC138526.1 | - | 18, 19 | 2247 | 8861546 | ||
| BC138527.1 | - | 18, 19 | 2247 | 8861547 | ||

Psmd1
Genomic Map