siRNA Details
Gene Symbol: Cenpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028960.1 | - | 1 | 1289 | ||
AK154611.1 | - | 4 | 2996 | |||
BC060191.1 | - | 5 | 3062 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cenpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028960.1 | - | 1 | 1289 | ||
AK154611.1 | - | 4 | 2996 | |||
BC060191.1 | - | 5 | 3062 |