siRNA Details
| Gene Symbol: 4921506M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037743.1 | NP_001032832.1 | 3 | 1109 | ||
| GenBank mRNA | AK014828.1 | - | 3 | 1100 | ||
| BC100489.1 | - | 3 | 1109 | 30915676 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921506M07Rik| Gene Symbol: 4921506M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037743.1 | NP_001032832.1 | 3 | 1109 | ||
| GenBank mRNA | AK014828.1 | - | 3 | 1100 | ||
| BC100489.1 | - | 3 | 1109 | 30915676 | ||