siRNA Details
Gene Symbol: 4931440P22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136269.1 | - | 2 | 1723 | ||
AK170145.1 | - | 2 | 1674 | |||
AK171113.1 | - | 1 | 1579 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_001033527.1 | - | 1674 | Chr. - - [-] on Build |
