siRNA Details
| Gene Symbol: Zfp597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033159.2 | NP_001028331.2 | 3 | 4421 | ||
| GenBank mRNA | AK042695.1 | - | 1 | 1960 | ||
| AK172006.1 | - | 2 | 1157 | |||
| BC057047.1 | - | 1 | 3830 | |||
| BC108421.1 | - | 1 | 3857 | |||

Zfp597
Genomic Map