siRNA Details
| Gene Symbol: Zfp597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033159.2 | NP_001028331.2 | 3 | 4626 | ||
| GenBank mRNA | AK042695.1 | - | 1 | 2165 | ||
| AK172006.1 | - | 2 | 1362 | |||
| BC057047.1 | - | 1 | 4035 | |||
| BC108421.1 | - | 1 | 4062 | |||

Zfp597
Genomic Map