siRNA Details
| Gene Symbol: 2700094K13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033166.2 | NP_001028338.1 | 4 | 597 | ||
| NM_001037279.1 | NP_001032356.1 | 4 | 572 | |||
| GenBank mRNA | AK012609.1 | - | 4 | 550 | ||
| AK167638.1 | - | 4 | 569 | |||
| AK171972.1 | - | 4 | 588 | |||
| BC056177.1 | - | 4 | 590 | 5683334 | ||
| CA318611.1 | - | 4 | 494 | |||

2700094K13Rik
Genomic Map