siRNA Details
Gene Symbol: 1700049G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028538.1 | NP_082814.1 | 4 | 3419 | ||
GenBank mRNA | AK137340.1 | - | 4 | 3419 | ||
BC141373.1 | - | 4 | 3412 | 9055986 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700049G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028538.1 | NP_082814.1 | 4 | 3419 | ||
GenBank mRNA | AK137340.1 | - | 4 | 3419 | ||
BC141373.1 | - | 4 | 3412 | 9055986 |