siRNA Details
| Gene Symbol: Wdr75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028599.2 | NP_082875.1 | 12 | 1154 | ||
| GenBank mRNA | AK010772.1 | - | 12 | 1154 | ||
| AK152412.1 | - | 12 | 1155 | |||
| AK163221.1 | - | 12 | 1128 | |||
| AK168032.1 | - | 12 | 1148 | |||
| AK168163.1 | - | 12 | 1156 | |||
| AK169932.1 | - | 12 | 1152 | |||
| BC055483.1 | - | 1 | 55 | |||
| BC085177.1 | - | 12 | 1184 | |||
| BC117880.2 | - | 12 | 1184 | 40096010 | ||
| BC117881.1 | - | 12 | 1181 | 40096014 | ||

Wdr75
Genomic Map