siRNA Details
| Gene Symbol: Klrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033171.2 | NP_001028343.2 | 5 | 2126 | ||
| GenBank mRNA | AK145064.1 | - | 5 | 2121 | ||
| AK162933.1 | - | 6 | 1716 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klrg2| Gene Symbol: Klrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033171.2 | NP_001028343.2 | 5 | 2126 | ||
| GenBank mRNA | AK145064.1 | - | 5 | 2121 | ||
| AK162933.1 | - | 6 | 1716 | |||