siRNA Details
| Gene Symbol: Klrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033171.2 | NP_001028343.2 | 5 | 1661 | ||
| GenBank mRNA | AK009417.1 | - | 5 | 1662 | ||
| AK145064.1 | - | 5 | 1656 | |||
| AK162933.1 | - | 6 | 1251 | |||
| AK169862.1 | - | 6 | 1205 | |||

Klrg2
Genomic Map