siRNA Details
| Gene Symbol: Ppapdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028922.3 | NP_083198.1 | 1 | 2057 | ||
| GenBank mRNA | AK016572.1 | - | 1 | 2062 | ||
| BC052412.1 | - | 1 | 2024 | 5720146 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppapdc2| Gene Symbol: Ppapdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028922.3 | NP_083198.1 | 1 | 2057 | ||
| GenBank mRNA | AK016572.1 | - | 1 | 2062 | ||
| BC052412.1 | - | 1 | 2024 | 5720146 | ||