siRNA Details
| Gene Symbol: Hyal5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028957.2 | NP_083233.1 | 4 | 1868 | ||
| GenBank mRNA | AK133387.1 | - | 4 | 1871 | ||
| BC145992.1 | - | 4 | 1801 | 40131295 | ||
| BC145994.1 | - | 4 | 1801 | 40131297 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hyal5| Gene Symbol: Hyal5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028957.2 | NP_083233.1 | 4 | 1868 | ||
| GenBank mRNA | AK133387.1 | - | 4 | 1871 | ||
| BC145992.1 | - | 4 | 1801 | 40131295 | ||
| BC145994.1 | - | 4 | 1801 | 40131297 | ||