siRNA Details
| Gene Symbol: CNNM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020348.2 | NP_065081.2 | 11 | 4121 | ||
| XM_005269692.3 | XP_005269749.2 | 10 | 3971 | |||
| XM_011539629.1 | XP_011537931.1 | 12 | 4184 | |||
| XM_011539630.1 | XP_011537932.1 | 11 | 4034 | |||
| GenBank mRNA | AF169226.1 | - | 11 | 2944 | ||
| AK056194.1 | - | 11 | 1735 | |||
| AL137536.1 | - | 4 | 1490 | |||
| BI868774.1 | - | 1 | 23 | |||

CNNM1
Genomic Map