siRNA Details
| Gene Symbol: Fam35a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029389.2 | NP_083665.2 | 4 | 1646 | ||
| GenBank mRNA | AK149442.1 | - | 4 | 1510 | ||
| AK162134.1 | - | 2 | 1456 | |||
| BC144822.1 | - | 1 | 1154 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam35a| Gene Symbol: Fam35a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029389.2 | NP_083665.2 | 4 | 1646 | ||
| GenBank mRNA | AK149442.1 | - | 4 | 1510 | ||
| AK162134.1 | - | 2 | 1456 | |||
| BC144822.1 | - | 1 | 1154 | |||