siRNA Details
| Gene Symbol: Fam35a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029389.2 | NP_083665.2 | 10 | 2807 | ||
| GenBank mRNA | AI451789.1 | - | 3 | 276 | ||
| AK149442.1 | - | 10 | 2671 | |||
| BC144822.1 | - | 7 | 2315 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam35a| Gene Symbol: Fam35a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029389.2 | NP_083665.2 | 10 | 2807 | ||
| GenBank mRNA | AI451789.1 | - | 3 | 276 | ||
| AK149442.1 | - | 10 | 2671 | |||
| BC144822.1 | - | 7 | 2315 | |||