siRNA Details
Gene Symbol: Tmem181a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033178.3 | NP_001028350.1 | 13 | 1206 | ||
GenBank mRNA | AK122507.1 | - | 10 | 2474 | ||
AK154554.1 | - | 13 | 1202 | |||
AK208144.1 | - | 1 | 44 | |||
BE862876.1 | - | 5 | 379 |
Gene Symbol: Tmem181b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039520.1 | - | 5 | 606 | ||
AK137412.1 | - | 6 | 1980 | |||
BC032285.1 | - | 5 | 1861 | |||
BC147312.1 | - | 1186 | 9056375 | |||
BC147313.1 | - | 1186 | 9056366 |
Gene Symbol: Tmem181c-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045744.1 | - | 5 | 869 |
