siRNA Details
| Gene Symbol: Itpripl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033380.3 | NP_001028552.1 | 1 | 5308 | ||
| GenBank mRNA | AK086993.1 | - | 1 | 2779 | ||
| AK134593.1 | - | 1 | 5256 | |||
| AK137678.1 | - | 1 | 5310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpripl2| Gene Symbol: Itpripl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033380.3 | NP_001028552.1 | 1 | 5308 | ||
| GenBank mRNA | AK086993.1 | - | 1 | 2779 | ||
| AK134593.1 | - | 1 | 5256 | |||
| AK137678.1 | - | 1 | 5310 | |||