siRNA Details
| Gene Symbol: Ripply1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037915.2 | NP_001033004.1 | 4 | 434 | ||
| GenBank mRNA | AB212222.1 | - | 4 | 446 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ripply1| Gene Symbol: Ripply1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037915.2 | NP_001033004.1 | 4 | 434 | ||
| GenBank mRNA | AB212222.1 | - | 4 | 446 | ||