siRNA Details
| Gene Symbol: Gm6460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037919.3 | NP_001033008.1 | 4 | 544 | ||
| GenBank mRNA | BC110565.1 | - | 4 | 544 | 40050213 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6460| Gene Symbol: Gm6460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037919.3 | NP_001033008.1 | 4 | 544 | ||
| GenBank mRNA | BC110565.1 | - | 4 | 544 | 40050213 | |