siRNA Details
Gene Symbol: Wdr93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037927.1 | NP_001033016.1 | 17 | 1945 | ||
GenBank mRNA | AB205021.1 | - | 17 | 1945 | ||
BC150920.1 | - | 16 | 1915 | 9087831 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wdr93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037927.1 | NP_001033016.1 | 17 | 1945 | ||
GenBank mRNA | AB205021.1 | - | 17 | 1945 | ||
BC150920.1 | - | 16 | 1915 | 9087831 |