siRNA Details
| Gene Symbol: Larp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138593.2 | NP_613059.2 | 3 | 404 | ||
| GenBank mRNA | AK012772.1 | - | 3 | 394 | ||
| AK015041.1 | - | 3 | 379 | |||
| AK082815.1 | - | 3 | 403 | |||
| AK162147.1 | - | 3 | 321 | |||
| BC020127.1 | - | 2 | 255 | |||
| BC023165.1 | - | 3 | 358 | |||
| BY269910.1 | - | 3 | 404 | |||

Larp7
Genomic Map