siRNA Details
| Gene Symbol: PLSCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199978.1 | NP_001186907.1 | 10 | 1451 | ||
| NM_001199979.1 | NP_001186908.1 | 8 | 1068 | |||
| NM_020359.2 | NP_065092.1 | 9 | 1181 | |||
| XM_017006910.1 | XP_016862399.1 | 8 | 1258 | |||
| XM_017006911.1 | XP_016862400.1 | 9 | 1535 | |||
| GenBank mRNA | AF159441.1 | - | 9 | 1125 | ||
| BC055415.1 | - | 8 | 1072 | |||
| BC069785.1 | - | 8 | 1018 | 7262547 | ||

PLSCR2
Genomic Map