siRNA Details
Gene Symbol: DMWD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004943.1 | NP_004934.1 | 5 | 2556 | ||
GenBank mRNA | BX283745.1 | - | 1 | 337 | ||
L19267.1 | - | 5 | 2146 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DMWD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004943.1 | NP_004934.1 | 5 | 2556 | ||
GenBank mRNA | BX283745.1 | - | 1 | 337 | ||
L19267.1 | - | 5 | 2146 |