siRNA Details
| Gene Symbol: HBZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005332.2 | NP_005323.1 | 3 | 571 | ||
| XM_005255287.3 | XP_005255344.1 | 4 | 1201 | |||
| XM_005255288.3 | XP_005255345.1 | 4 | 1318 | |||
| GenBank mRNA | BC027892.1 | - | 3 | 551 | 5224569 | |

HBZ
Genomic Map