siRNA Details
| Gene Symbol: PITHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020362.4 | NP_065095.2 | 4 | 541 | ||
| XM_011541804.1 | XP_011540106.1 | 3 | 466 | |||
| GenBank mRNA | AF218024.1 | - | 4 | 522 | ||
| AF221595.1 | - | 4 | 533 | |||
| AF271784.1 | - | 4 | 537 | |||
| AK296662.1 | - | 3 | 466 | |||
| AK310330.1 | - | 4 | 488 | |||
| AK313005.1 | - | 4 | 509 | |||
| BC017208.2 | - | 4 | 502 | 3943360 | ||

PITHD1
Genomic Map