siRNA Details
| Gene Symbol: PGAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY007118.1 | - | 1 | 1695 | ||
| BC000455.1 | - | 1 | 1721 | 2821567 | ||
| BC010038.1 | - | 1 | 1713 | 3604026 | ||
| BC011678.2 | - | 1 | 1677 | 4299008 | ||
| BC053356.1 | - | 1 | 1694 | 5787678 | ||
| BC073742.1 | - | 1 | 1692 | 6297173 | ||

PGAM1
Genomic Map