siRNA Details
| Gene Symbol: PSG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113410.1 | NP_001106881.1 | 3 | 495 | ||
| NM_002785.2 | NP_002776.3 | 4 | 861 | |||
| NM_203287.1 | NP_976032.2 | 3 | 495 | |||
| GenBank mRNA | BC020711.1 | - | 3 | 485 | 4732286 | |
| BG571293.1 | - | 2 | 204 | |||
| M69245.1 | - | 4 | 823 | |||
| R66665.1 | - | 2 | 209 | |||
| U25988.1 | - | 3 | 491 | |||

PSG11
Genomic Map