siRNA Details
| Gene Symbol: SCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021920.3 | NP_068739.1 | 4 | 561 | ||
| XM_017018109.1 | XP_016873598.1 | 4 | 557 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCT| Gene Symbol: SCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021920.3 | NP_068739.1 | 4 | 561 | ||
| XM_017018109.1 | XP_016873598.1 | 4 | 557 | |||