siRNA Details
| Gene Symbol: SPRR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017418.2 | NP_001017418.1 | 2 | 377 | ||
| XM_017002174.1 | XP_016857663.1 | 2 | 373 | |||
| GenBank mRNA | BF090370.1 | - | 1 | 249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRR2B| Gene Symbol: SPRR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017418.2 | NP_001017418.1 | 2 | 377 | ||
| XM_017002174.1 | XP_016857663.1 | 2 | 373 | |||
| GenBank mRNA | BF090370.1 | - | 1 | 249 | ||