siRNA Details
| Gene Symbol: TPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286272.1 | NP_001273201.1 | 4 | 524 | ||
| NM_001286273.1 | NP_001273202.1 | 3 | 450 | |||
| NM_003295.3 | NP_003286.1 | 4 | 524 | |||
| GenBank mRNA | AK130847.1 | - | 4 | 423 | ||
| AK312951.1 | - | 4 | 423 | |||
| AY117678.1 | - | 4 | 424 | |||
| AY334563.1 | - | 4 | 309 | |||
| BC003352.2 | - | 4 | 407 | 2899964 | ||
| BC012431.1 | - | 4 | 383 | 3880559 | ||
| BC022436.1 | - | 3 | 378 | 4281364 | ||
| BC040008.1 | - | 2 | 711 | |||
| BC052333.1 | - | 4 | 413 | 6058799 | ||
| BP215975.1 | - | 4 | 525 | |||
| CR457036.1 | - | 4 | 309 | |||
| L13806.1 | - | 2 | 144 | |||
| X16064.1 | - | 4 | 403 | |||
| X64899.1 | - | 4 | 395 | |||

TPT1
Genomic Map