siRNA Details
| Gene Symbol: HCG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028032.1 | 3 | 525 | |||
| GenBank mRNA | X95289.1 | - | 3 | 520 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCG9| Gene Symbol: HCG9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028032.1 | 3 | 525 | |||
| GenBank mRNA | X95289.1 | - | 3 | 520 | ||