siRNA Details
| Gene Symbol: CLDN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171092.1 | NP_001164563.1 | 2 | 1013 | ||
| NM_001171095.1 | NP_001164566.1 | 2 | 782 | |||
| NM_020384.3 | NP_065117.1 | 2 | 848 | |||
| GenBank mRNA | AF177340.1 | - | 2 | 1013 | ||
| AF250558.1 | - | 1 | 494 | |||
| AK075371.1 | - | 2 | 724 | |||
| AK075405.1 | - | 2 | 820 | |||
| AK312515.1 | - | 2 | 782 | |||
| AY358474.1 | - | 1 | 615 | |||
| BC014424.1 | - | 1 | 615 | 4645075 | ||
| BC071747.1 | - | 2 | 821 | 30322852 | ||

CLDN2
Genomic Map