siRNA Details
Gene Symbol: SOGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022023.1 | - | 1 | 1877 | ||
BC113405.1 | - | 1 | 1637 | |||
BC113433.1 | - | 1 | 1637 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SOGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK022023.1 | - | 1 | 1877 | ||
BC113405.1 | - | 1 | 1637 | |||
BC113433.1 | - | 1 | 1637 |