siRNA Details
| Gene Symbol: PURG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015508.2 | NP_001015508.1 | 1 | 1617 | ||
| NM_001323311.1 | NP_001310240.1 | 2 | 807 | |||
| NM_001323312.1 | NP_001310241.1 | 2 | 807 | |||
| NM_013357.2 | NP_037489.1 | 1 | 703 | |||
| GenBank mRNA | AF195513.2 | - | 1 | 1617 | ||
| AY077841.1 | - | 1 | 1617 | |||
| BC106708.2 | - | 1 | 703 | 40005076 | ||

PURG
Genomic Map