siRNA Details
| Gene Symbol: STAG3L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040582.2 | 9 | 1713 | |||
| GenBank mRNA | BC030509.1 | - | 5 | 990 | ||
| Gene Symbol: STAG3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040583.2 | 10 | 1694 | |||
| Gene Symbol: STAG3L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040584.2 | 8 | 1438 | |||
| GenBank mRNA | BM311130.1 | - | 1 | 60 | ||

STAG3L3
Genomic Map