siRNA Details
| Gene Symbol: PGLYRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020393.3 | NP_065126.2 | 9 | 2022 | ||
| XM_011509789.2 | XP_011508091.1 | 9 | 2034 | |||
| XM_011509791.2 | XP_011508093.1 | 9 | 2013 | |||
| XM_011509792.1 | XP_011508094.1 | 9 | 1714 | |||
| XM_011509793.2 | XP_011508095.1 | 6 | 1710 | |||
| GenBank mRNA | AF086392.1 | - | 1 | 344 | ||
| AF242518.1 | - | 9 | 1732 | |||

PGLYRP4
Genomic Map