siRNA Details
| Gene Symbol: LY6G6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021246.2 | NP_067069.2 | 2 | 169 | ||
| GenBank mRNA | AJ315535.1 | - | 2 | 169 | ||
| AJ315536.1 | - | 1 | 260 | |||
| BC125138.1 | - | 2 | 173 | 40119583 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LY6G6D| Gene Symbol: LY6G6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021246.2 | NP_067069.2 | 2 | 169 | ||
| GenBank mRNA | AJ315535.1 | - | 2 | 169 | ||
| AJ315536.1 | - | 1 | 260 | |||
| BC125138.1 | - | 2 | 173 | 40119583 | ||